accessibility menu, dialog, popup

UserName

CAS RN 87749-50-6

IUPAC Name: tetrabutylazanium trihydrate fluoride
Synonyms: tetrabutylammonium ion trihydrate fluoride
Molecular Weight (g/mol): 315.51
Molecular Formula: C16H42FNO3
InChi Key: VEPTXBCIDSFGBF-UHFFFAOYSA-M
SMILES: O.O.O.[F-].CCCC[N+](CCCC)(CCCC)CCCC
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (5)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (3)