accessibility menu, dialog, popup

UserName

CH3H3CCH3OH

CAS RN 877-65-6

IUPAC Name: (4-tert-butylphenyl)methanol
Synonyms: 4-tert-butylbenzyl alcohol
Molecular Weight (g/mol): 164.25
Molecular Formula: C11H16O
InChi Key: FVEINXLJOJPHLH-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC=C(CO)C=C1
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)