CAS RN 876-31-3
IUPAC Name:
4-(cyanomethyl)benzonitrile
Synonyms:
4-(cyanomethyl)benzonitrile
Molecular Weight (g/mol):
142.16
Molecular Formula:
C9H6N2
InChi Key:
QILKKAFYAFEWGU-UHFFFAOYSA-N
SMILES:
N#CCC1=CC=C(C=C1)C#N
1
–
1
of
1
results
1
4-Cyanophenylacetonitrile, 97%
CAS: 876-31-3 Molecular Formula: C9H6N2 Molecular Weight (g/mol): 142.16 MDL Number: MFCD00060305 InChI Key: QILKKAFYAFEWGU-UHFFFAOYSA-N Synonym: 4-cyanomethyl benzonitrile,4-cyanophenylacetonitrile,benzeneacetonitrile, 4-cyano,p-tolunitrile, .alpha.-cyano,p-cyanobenzylcyanide,4-cyanobenzeneacetonitrile,.alpha.-cyano-p-tolunitrile,4-cyanbenzylcyanide,4-cyanobenzylcyanide,acmc-209qoz PubChem CID: 136686 IUPAC Name: 4-(cyanomethyl)benzonitrile SMILES: N#CCC1=CC=C(C=C1)C#N