accessibility menu, dialog, popup

UserName

CAS RN 87508-17-6

OSOHOHOO

CAS RN 87508-17-6

IUPAC Name: 2-phenyl-6-(phenylsulfanyl)-hexahydro-2H-pyrano[3,2-d][1,3]dioxine-7,8-diol
Synonyms: 2-phenyl-6-(phenylsulfanyl)-hexahydro-2H-pyrano[3,2-d][1,3]dioxine-7,8-diol
Molecular Weight (g/mol): 360.42
Molecular Formula: C19H20O5S
InChi Key: BDNIQCYVYFGHSI-UHFFFAOYNA-N
SMILES: OC1C(O)C2OC(OCC2OC1SC1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results