CAS RN 874-66-8
IUPAC Name:
(2Z)-3-(furan-2-yl)-2-methylprop-2-enal
Synonyms:
(2Z)-3-(furan-2-yl)-2-methylprop-2-enal
Molecular Weight (g/mol):
136.15
Molecular Formula:
C8H8O2
InChi Key:
ZNBXZUKDRRRQJK-ALCCZGGFSA-N
SMILES:
C\C(C=O)=C\C1=CC=CO1
1
–
1
of
1
results
1
Promotions Available
2-Methyl-3-(2-furyl)propenal, 97%
2-Methyl-3-(2-furyl)propenal, 97%
CAS: 874-66-8 Molecular Formula: C8H8O2 Molecular Weight (g/mol): 136.15 MDL Number: MFCD00063240 InChI Key: ZNBXZUKDRRRQJK-FNORWQNLSA-N Synonym: 2-methyl-3-2-furyl propenal,alpha-methylfurylacrolein,2-methyl-3-2-furyl acrolein,furfurylidene-2-propanal,furfurylidine-2-propanal,2-methyl-3-furylacrolein,alpha-methyl-2-furanacrolein,2-furfurylidenepropionaldehyde,3-2-furyl methacrylaldehyde,2-methyl-3 2-furyl acrolein PubChem CID: 6210220 IUPAC Name: (E)-3-(furan-2-yl)-2-methylprop-2-enal SMILES: CC(=CC1=CC=CO1)C=O