accessibility menu, dialog, popup

UserName

CAS RN 873-64-3

CH3NSH3CCH3

CAS RN 873-64-3

IUPAC Name: 2-ethyl-4,5-dimethyl-1,3-thiazole
Synonyms: 2-ethyl-4,5-dimethyl-1,3-thiazole
Molecular Weight (g/mol): 141.23
Molecular Formula: C7H11NS
InChi Key: BVOKJOZBJOWZNJ-UHFFFAOYSA-N
SMILES: CCC1=NC(C)=C(C)S1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results