accessibility menu, dialog, popup

UserName

CAS RN 872-10-6

IUPAC Name: 1-(pentylsulfanyl)pentane
Synonyms: diamyl sulfide
Molecular Weight (g/mol): 174.35
Molecular Formula: C10H22S
InChi Key: JOZDADPMWLVEJK-UHFFFAOYSA-N
SMILES: CCCCCSCCCCC
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)