accessibility menu, dialog, popup

UserName

CAS RN 87199-18-6

IUPAC Name: (3-hydroxyphenyl)boronic acid
Synonyms: 3-hydroxyphenylboronic acid
Molecular Weight (g/mol): 137.93
Molecular Formula: C6H7BO3
InChi Key: WFWQWTPAPNEOFE-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=CC(O)=C1
Quantity 
  • (3)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (4)
  • (1)
Physical Form 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (5)
  • (3)
  • (1)
  • (1)
  • (1)
  • (4)
  • (1)
  • (1)
  • (2)