missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxybenzeneboronic acid, 97%

Catalog No. AAL1906106
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1906106 5 g
AAL1906103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1906106 Supplier Thermo Scientific Chemicals Supplier No. L1906106
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 87199-18-6 | C6H7BO3 | 137.93 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 87199-18-6
Molecular Formula C6H7BO3
Molecular Weight (g/mol) 137.93
MDL Number MFCD01074603
InChI Key WFWQWTPAPNEOFE-UHFFFAOYSA-N
Synonym 3-hydroxyphenyl boronic acid, 3-hydroxybenzeneboronic acid, 3-hydroxyphenylboronicacid, 3-hydroxyphenyl boranediol, m-hydroxyphenylboronic acid, boronic acid, 3-hydroxyphenyl, pubchem1728, 3-boronophenol, acmc-209qip, 3-hydroxyphenyboronic acid
PubChem CID 2734359
IUPAC Name (3-hydroxyphenyl)boronic acid
SMILES OB(O)C1=CC=CC(O)=C1

Specifications

Melting Point 208°C to 210°C
Quantity 5 g
Beilstein 3240771
Formula Weight 137.93
Percent Purity 97%
Chemical Name or Material 3-Hydroxybenzeneboronic acid
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only