missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxyphenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. H11851G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H11851G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H11851G Supplier TCI America Supplier No. H11851G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material 3-Hydroxyphenylboronic Acid (contains varying amounts of Anhydride)
CAS 87199-18-6
Melting Point 213°C
Molecular Formula C6H7BO3
Quantity 1 g
MDL Number MFCD01074603
Synonym 3-hydroxyphenyl boronic acid, 3-hydroxybenzeneboronic acid, 3-hydroxyphenylboronicacid, 3-hydroxyphenyl boranediol, m-hydroxyphenylboronic acid, boronic acid, 3-hydroxyphenyl, pubchem1728, 3-boronophenol, acmc-209qip, 3-hydroxyphenyboronic acid
InChI Key WFWQWTPAPNEOFE-UHFFFAOYSA-N
SMILES OB(O)C1=CC=CC(O)=C1
IUPAC Name (3-hydroxyphenyl)boronic acid
Molecular Weight (g/mol) 137.93
PubChem CID 2734359
Formula Weight 137.93
Physical Form Crystalline Powder
Show More Show Less