CAS RN 871696-12-7
CAS RN 871696-12-7
IUPAC Name:
9-hexyl-2,7-bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9H-carbazole
Synonyms:
9-hexyl-2,7-bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)carbazole
Molecular Weight (g/mol):
503.3
Molecular Formula:
C30H43B2NO4
InChi Key:
VUJPSYQJTJSPOI-UHFFFAOYSA-N
SMILES:
CCCCCCN1C2=C(C=CC(=C2)B2OC(C)(C)C(C)(C)O2)C2=C1C=C(C=C2)B1OC(C)(C)C(C)(C)O1
1
–
1
of
1
results
1
9-Hexyl-2,7-bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9H-carbazole 98.0+%, TCI America™
CAS: 871696-12-7 Molecular Formula: C30H43B2NO4 Molecular Weight (g/mol): 503.297 MDL Number: MFCD30187269 InChI Key: VUJPSYQJTJSPOI-UHFFFAOYSA-N PubChem CID: 25150161 IUPAC Name: 9-hexyl-2,7-bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)carbazole SMILES: B1(OC(C(O1)(C)C)(C)C)C2=CC3=C(C=C2)C4=C(N3CCCCCC)C=C(C=C4)B5OC(C(O5)(C)C)(C)C