CAS RN 87151-66-4
IUPAC Name:
3-(cyclohexa-1,4-dien-1-yl)propan-1-ol
Synonyms:
3-(cyclohexa-1,4-dien-1-yl)propan-1-ol
Molecular Weight (g/mol):
138.21
Molecular Formula:
C9H14O
InChi Key:
DJJNAQALDCPMOU-UHFFFAOYSA-N
SMILES:
OCCCC1=CCC=CC1
1
–
1
of
1
results
1
Promotions Available
3-(1,4-Cyclohexadien-1-yl)-1-propanol, 97%
3-(1,4-Cyclohexadien-1-yl)-1-propanol, 97%
CAS: 87151-66-4 Molecular Formula: C9H14O Molecular Weight (g/mol): 138.21 MDL Number: MFCD12407081 InChI Key: DJJNAQALDCPMOU-UHFFFAOYSA-N Synonym: 3-1,4-cyclohexadien-1-yl-1-propanol,3-cyclohexa-1,4-dien-1-yl propan-1-ol,3-1,4-cyclohexadienyl propan-1-ol,3-3-hydroxypropyl-1,4-cyclohexadiene PubChem CID: 20402847 IUPAC Name: 3-cyclohexa-1,4-dien-1-ylpropan-1-ol SMILES: C1C=CCC(=C1)CCCO