CAS RN 871332-96-6
IUPAC Name:
[3-ethoxy-5-(trifluoromethyl)phenyl]boronic acid
Synonyms:
3-ethoxy-5-(trifluoromethyl)phenylboronic acid
Molecular Weight (g/mol):
233.98
Molecular Formula:
C9H10BF3O3
InChi Key:
ATZRXKHRSOTBAM-UHFFFAOYSA-N
SMILES:
CCOC1=CC(=CC(=C1)C(F)(F)F)B(O)O
1
–
1
of
1
results
1
Promotions Available
3-Ethoxy-5-(trifluoromethyl)benzeneboronic acid, 98%, Thermo Scientific Chemicals
3-Ethoxy-5-(trifluoromethyl)benzeneboronic acid, 98%, Thermo Scientific Chemicals
CAS: 871332-96-6 Molecular Formula: C9H10BF3O3 Molecular Weight (g/mol): 233.98 MDL Number: MFCD08056361 InChI Key: ATZRXKHRSOTBAM-UHFFFAOYSA-N Synonym: 3-ethoxy-5-trifluoromethyl phenylboronic acid,3-ethoxy-5-trifluoromethyl benzeneboronic acid,boronic acid,b-3-ethoxy-5-trifluoromethyl phenyl,3-ethoxy-5-trifluoromethylphenylboronic acid,3-ethoxy-5-trifluoromethyl phenyl boronic acid,acmc-209qhp,3-ethoxy-5-trifluoromethyl benzeneboronicacid PubChem CID: 44717589 IUPAC Name: [3-ethoxy-5-(trifluoromethyl)phenyl]boronic acid SMILES: CCOC1=CC(=CC(=C1)C(F)(F)F)B(O)O