accessibility menu, dialog, popup

UserName

CH3ONH2NOHO

CAS RN 870997-81-2

IUPAC Name: 3-amino-2-methoxypyridine-4-carboxylic acid
Synonyms: 3-amino-2-methoxypyridine-4-carboxylic acid
Molecular Weight (g/mol): 168.15
Molecular Formula: C7H8N2O3
InChi Key: ABVDQGNEDSKBLA-UHFFFAOYSA-N
SMILES: COC1=NC=CC(C(O)=O)=C1N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results