accessibility menu, dialog, popup

UserName

CAS RN 870483-87-7

H3COONNNH2H2NOCH3

CAS RN 870483-87-7

IUPAC Name: 5-({3-methoxy-4-[(4-methoxyphenyl)methoxy]phenyl}methyl)pyrimidine-2,4-diamine
Synonyms: 5-({3-methoxy-4-[(4-methoxyphenyl)methoxy]phenyl}methyl)pyrimidine-2,4-diamine
Molecular Weight (g/mol): 366.42
Molecular Formula: C20H22N4O3
InChi Key: MYQAUKPBNJWPIE-UHFFFAOYSA-N
SMILES: COC1=CC=C(COC2=C(OC)C=C(CC3=CN=C(N)N=C3N)C=C2)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results