accessibility menu, dialog, popup

UserName

CH3H3CCH3OOHO

CAS RN 87-18-3

IUPAC Name: 4-tert-butylphenyl 2-hydroxybenzoate
Synonyms: 4-tert-butylphenyl 2-hydroxybenzoate
Molecular Weight (g/mol): 270.33
Molecular Formula: C17H18O3
InChi Key: DBOSBRHMHBENLP-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC=C(OC(=O)C2=CC=CC=C2O)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results