accessibility menu, dialog, popup

UserName

CAS RN 87-02-5

H2NOHSOHOO

CAS RN 87-02-5

IUPAC Name: 7-amino-4-hydroxynaphthalene-2-sulfonic acid
Synonyms: 7-amino-4-hydroxynaphthalene-2-sulfonic acid
Molecular Weight (g/mol): 239.25
Molecular Formula: C10H9NO4S
InChi Key: KYARBIJYVGJZLB-UHFFFAOYSA-N
SMILES: NC1=CC=C2C(O)=CC(=CC2=C1)S(O)(=O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)