accessibility menu, dialog, popup

UserName

CAS RN 86891-03-4

IUPAC Name: sodium 4-oxopent-2-en-2-olate
Synonyms: sodium 4-oxopent-2-en-2-olate
Molecular Weight (g/mol): 122.1
Molecular Formula: C5H7NaO2
InChi Key: AIWZOHBYSFSQGV-UHFFFAOYSA-M
SMILES: [Na+].CC([O-])=CC(C)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results