accessibility menu, dialog, popup

UserName

CH3H3CCH3ONHN

CAS RN 86847-59-8

IUPAC Name: 2,2-dimethyl-N-(pyridin-2-yl)propanamide
Synonyms: 2,2-dimethyl-N-(pyridin-2-yl)propanamide
Molecular Weight (g/mol): 178.24
Molecular Formula: C10H14N2O
InChi Key: CGSPVYCZBDFPHJ-UHFFFAOYSA-N
SMILES: CC(C)(C)C(=O)NC1=CC=CC=N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results