accessibility menu, dialog, popup

UserName

H3CCH2OOOH

CAS RN 868-77-9

IUPAC Name: 2-hydroxyethyl 2-methylprop-2-enoate
Synonyms: HEMA monomer MG-1 polyhydroxyethylmethacrylate mhoromer (hydroxyethyl)methacrylate monomethacrylic ether of ethylene glycol 2-hydroxyethylmethacrylate
Molecular Weight (g/mol): 130.14
Molecular Formula: C6H10O3
InChi Key: WOBHKFSMXKNTIM-UHFFFAOYSA-N
SMILES: CC(=C)C(=O)OCCO
Boiling Point 
  • (4)
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (4)
  • (4)
Percent Purity 
  • (2)
  • (6)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (6)
  • (4)
  • (2)
  • (2)
  • (4)
  • (4)
  • (2)
  • (6)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (6)