accessibility menu, dialog, popup

UserName

H3COOCH3SiCH3H3CCH3OSOOFFF

CAS RN 866252-52-0

IUPAC Name: 4,5-dimethoxy-2-(trimethylsilyl)phenyl trifluoromethanesulfonate
Synonyms: 4,5-dimethoxy-2-(trimethylsilyl)phenyl trifluoromethanesulfonate
Molecular Weight (g/mol): 358.4
Molecular Formula: C12H17F3O5SSi
InChi Key: DLUVZOFWZNLGTJ-UHFFFAOYSA-N
SMILES: COC1=CC(OS(=O)(=O)C(F)(F)F)=C(C=C1OC)[Si](C)(C)C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results