accessibility menu, dialog, popup

UserName

CH3H3COBOH3CH3CNNS

CAS RN 864754-19-8

IUPAC Name: 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1-[(thiophen-2-yl)methyl]-1H-pyrazole
Synonyms: 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1-(thiophen-2-ylmethyl)pyrazole
Molecular Weight (g/mol): 290.19
Molecular Formula: C14H19BN2O2S
InChi Key: ANSBFDYOIUZSNT-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CN(CC2=CC=CS2)N=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results