CAS RN 86267-86-9
IUPAC Name:
3,4-dihydro-2H-1,4-benzoxazine-2-carbonitrile
Synonyms:
3,4-dihydro-2H-1,4-benzoxazine-2-carbonitrile
Molecular Weight (g/mol):
160.18
Molecular Formula:
C9H8N2O
InChi Key:
YSTANLOUKDVPGJ-UHFFFAOYNA-N
SMILES:
N#CC1CNC2=CC=CC=C2O1
1
–
2
of
2
results
2
3,4-Dihydro-2H-1,4-benzoxazine-2-carbonitrile, 97+%, Thermo Scientific™
CAS: 86267-86-9 Molecular Formula: C9H8N2O Molecular Weight (g/mol): 160.176 InChI Key: YSTANLOUKDVPGJ-UHFFFAOYSA-N Synonym: 3,4-dihydro-2h-benzo b 1,4 oxazine-2-carbonitrile,2h,3h,4h-benzo e 1,4-oxazaperhydroine-2-carbonitrile,2h-1,4-benzoxazine-2-carbonitrile,3,4-dihydro PubChem CID: 2795504 IUPAC Name: 3,4-dihydro-2H-1,4-benzoxazine-2-carbonitrile SMILES: C1C(OC2=CC=CC=C2N1)C#N