accessibility menu, dialog, popup

UserName

CAS RN 86256-24-8

IUPAC Name: 2-(bromomethyl)-1-fluoro-4-(trifluoromethoxy)benzene
Synonyms: 2-(bromomethyl)-1-fluoro-4-(trifluoromethoxy)benzene
Molecular Weight (g/mol): 273.03
Molecular Formula: C8H5BrF4O
InChi Key: KOISGBFWQAPLFE-UHFFFAOYSA-N
SMILES: FC1=C(CBr)C=C(OC(F)(F)F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results