accessibility menu, dialog, popup

UserName

CAS RN 86116-84-9

IUPAC Name: hydrogen (2R)-2-benzylpyrrolidine-2-carboxylic acid chloride
Synonyms: hydrogen (2R)-2-benzylpyrrolidine-2-carboxylic acid chloride
Molecular Weight (g/mol): 241.72
Molecular Formula: C12H16ClNO2
InChi Key: SPHJFGQFCBZKRU-UTONKHPSSA-N
SMILES: [H+].[Cl-].OC(=O)[C@]1(CC2=CC=CC=C2)CCCN1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)