accessibility menu, dialog, popup

UserName

BrOOO

CAS RN 86-90-8

IUPAC Name: 5-bromo-1,3-dihydro-2-benzofuran-1,3-dione
Synonyms: 5-bromo-2-benzofuran-1,3-dione
Molecular Weight (g/mol): 227.01
Molecular Formula: C8H3BrO3
InChi Key: BCKVHOUUJMYIAN-UHFFFAOYSA-N
SMILES: BrC1=CC=C2C(=O)OC(=O)C2=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results