accessibility menu, dialog, popup

UserName

OHO

CAS RN 86-87-3

IUPAC Name: 2-(naphthalen-1-yl)acetic acid
Synonyms: plucker Tip-Off caswell no. 589 rasin acide naphthylacetique planofix liqui-stik primacol pomoxon stafast
Molecular Weight (g/mol): 186.21
Molecular Formula: C12H10O2
InChi Key: PRPINYUDVPFIRX-UHFFFAOYSA-N
SMILES: OC(=O)CC1=C2C=CC=CC2=CC=C1
Percent Purity 
  • (2)
  • (3)
  • (3)
  • (6)
Quantity 
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
Grade 
  • (6)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (12)
  • (6)
  • (2)
  • (6)
  • (2)
  • (3)
  • (3)
  • (6)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (6)
  • (2)
  • (3)