accessibility menu, dialog, popup

UserName

H2NHNNH2

CAS RN 86-71-5

IUPAC Name: 9H-carbazole-3,6-diamine
Synonyms: 9H-carbazole-3,6-diamine
Molecular Weight (g/mol): 197.24
Molecular Formula: C12H11N3
InChi Key: YCZUWQOJQGCZKG-UHFFFAOYSA-N
SMILES: NC1=CC2=C(NC3=C2C=C(N)C=C3)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results