accessibility menu, dialog, popup

UserName

BCH3NH3CCH3

CAS RN 85874-45-9

IUPAC Name: benzyltriethylazanium λ¹-boranuidylidene
Synonyms: benzyltriethylazanium λ¹-boranuidylidene
Molecular Weight (g/mol): 203.14
Molecular Formula: C13H22BN
InChi Key: HQGWCEJYXHFBSA-UHFFFAOYSA-N
SMILES: [B-].CC[N+](CC)(CC)CC1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results