accessibility menu, dialog, popup

UserName

CH3OOCH3CH3H2NHCl

CAS RN 85532-40-7

IUPAC Name: ethyl 3-amino-3-methylbutanoate hydrochloride
Synonyms: ethyl 3-amino-3-methylbutanoate hydrochloride
Molecular Weight (g/mol): 181.66
Molecular Formula: C7H16ClNO2
InChi Key: HFBJSIGXRKAYAF-UHFFFAOYSA-N
SMILES: Cl.CCOC(=O)CC(C)(C)N
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results