accessibility menu, dialog, popup

UserName

CAS RN 855-38-9

IUPAC Name: tris(4-methoxyphenyl)phosphane
Synonyms: tris(4-methoxyphenyl)phosphane
Molecular Weight (g/mol): 352.37
Molecular Formula: C21H21O3P
InChi Key: UYUUAUOYLFIRJG-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)P(C1=CC=C(OC)C=C1)C1=CC=C(OC)C=C1
Molecular Weight (g/mol) 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (4)