missing translation for 'onlineSavingsMsg'
Learn More

Tris(4-methoxyphenyl)phosphine 97.0+%, TCI America™

Catalog No. T08615G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T08615G 5 g
T086125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T08615G Supplier TCI America Supplier No. T08615G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 855-38-9
Molecular Formula C21H21O3P
Molecular Weight (g/mol) 352.37
MDL Number MFCD00014896
InChI Key UYUUAUOYLFIRJG-UHFFFAOYSA-N
Synonym tris 4-methoxyphenyl phosphine, trianisylphosphine, tris p-anisyl phosphine, phosphine, tris 4-methoxyphenyl, tri-p-anisylphosphine, tris p-methoxyphenyl phosphine, tris 4-methoxyphenyl phosphane, phosphine, tris p-methoxyphenyl, tri 4-methoxyphenyl phosphine, tri p-methoxyphenyl phosphine
PubChem CID 70071
IUPAC Name tris(4-methoxyphenyl)phosphane
SMILES COC1=CC=C(C=C1)P(C2=CC=C(C=C2)OC)C3=CC=C(C=C3)OC

Specifications

Melting Point 133°C
Color White-Yellow
Quantity 5 g
Formula Weight 352.37
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tris(4-methoxyphenyl)phosphine