missing translation for 'onlineSavingsMsg'
Learn More

Tris(4-methoxyphenyl)phosphine, 95%

Catalog No. AC422240100
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC422240100 10 g Glass bottle
AC422240010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC422240100 Supplier Thermo Scientific Chemicals Supplier No. 422240100
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 855-38-9 | C21H21O3P | 352.36 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 855-38-9
Molecular Formula C21H21O3P
Molecular Weight (g/mol) 352.36
MDL Number MFCD00014896
InChI Key UYUUAUOYLFIRJG-UHFFFAOYSA-N
Synonym tris 4-methoxyphenyl phosphine, trianisylphosphine, tris p-anisyl phosphine, phosphine, tris 4-methoxyphenyl, tri-p-anisylphosphine, tris p-methoxyphenyl phosphine, tris 4-methoxyphenyl phosphane, phosphine, tris p-methoxyphenyl, tri 4-methoxyphenyl phosphine, tri p-methoxyphenyl phosphine
PubChem CID 70071
IUPAC Name tris(4-methoxyphenyl)phosphane
SMILES COC1=CC=C(C=C1)P(C2=CC=C(C=C2)OC)C3=CC=C(C=C3)OC

Specifications

Melting Point 131.0°C to 134.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Quantity 10 g
Solubility Information Solubility in water: practically insoluble. Other solubilities: soluble in methanol
Formula Weight 352.36
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material Tris(4-methoxyphenyl)phosphine, 95%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/e
EINECSNumber 212-723-0
TSCA TSCA

RUO – Research Use Only