accessibility menu, dialog, popup

UserName

CAS RN 85483-97-2

IUPAC Name: 1-(benzyloxy)propan-2-ol
Synonyms: 1-(benzyloxy)propan-2-ol
Molecular Weight (g/mol): 166.22
Molecular Formula: C10H14O2
InChi Key: KJBPYIUAQLPHJG-UHFFFAOYNA-N
SMILES: CC(O)COCC1=CC=CC=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results