accessibility menu, dialog, popup

UserName

CAS RN 85416-75-7

H3COOHNO

CAS RN 85416-75-7

IUPAC Name: 4-[3-(cyclopentyloxy)-4-methoxyphenyl]pyrrolidin-2-one
Synonyms: rolipram bevacizumab 咯利普兰
Molecular Weight (g/mol): 275.35
Molecular Formula: C16H21NO3
InChi Key: HJORMJIFDVBMOB-UHFFFAOYNA-N
SMILES: COC1=CC=C(C=C1OC1CCCC1)C1CNC(=O)C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results