CAS RN 85416-73-5
CAS RN 85416-73-5
IUPAC Name:
(4S)-4-[3-(cyclopentyloxy)-4-methoxyphenyl]pyrrolidin-2-one
Synonyms:
(+)-rolipram (S)-(+)-rolipram
Molecular Weight (g/mol):
275.35
Molecular Formula:
C16H21NO3
InChi Key:
HJORMJIFDVBMOB-GFCCVEGCSA-N
SMILES:
COC1=CC=C(C=C1OC1CCCC1)[C@H]1CNC(=O)C1
1
–
1
of
1
results
1
(S)-(+)-Rolipram 98.0+%, TCI America™
CAS: 85416-73-5 Molecular Formula: C16H21NO3 Molecular Weight (g/mol): 275.348 MDL Number: MFCD03093861 InChI Key: HJORMJIFDVBMOB-GFCCVEGCSA-N Synonym: s-+-rolipram,s-rolipram,+-rolipram,unii-h6g4vmq24k,h6g4vmq24k,4s-4-3-cyclopentyloxy-4-methoxyphenyl pyrrolidin-2-one,s-4-3-cyclopentyloxy-4-methoxyphenyl pyrrolidin-2-one PubChem CID: 158758 ChEBI: CHEBI:59540 IUPAC Name: (4S)-4-(3-cyclopentyloxy-4-methoxyphenyl)pyrrolidin-2-one SMILES: COC1=C(C=C(C=C1)C2CC(=O)NC2)OC3CCCC3