CAS RN 85316-98-9
IUPAC Name:
N-methyl-N-[(2S,3R,4R,5R)-2,3,4,5,6-pentahydroxyhexyl]octanamide
Synonyms:
N-methyl-N-[(2S,3R,4R,5R)-2,3,4,5,6-pentahydroxyhexyl]octanamide
Molecular Weight (g/mol):
321.41
Molecular Formula:
C15H31NO6
InChi Key:
SBWGZAXBCCNRTM-CTHBEMJXSA-N
SMILES:
CCCCCCCC(=O)N(C)C[C@H](O)[C@@H](O)[C@H](O)[C@H](O)CO
1
–
1
of
1
results
1
Thermo Scientific Chemicals N-Octanoyl-N-methyl-D-glucamine
CAS: 85316-98-9 Molecular Formula: C15H31NO6 Molecular Weight (g/mol): 321.414 MDL Number: MFCD00134152 InChI Key: SBWGZAXBCCNRTM-CTHBEMJXSA-N Synonym: mega-8,n-octanoyl-n-methylglucamine,mega 8,mega-8 solution,n-methyl-n-octanoyl-d-glucamine,n-d-glucityl-n-methyloctanamide,mega-8 500 mm solution,n-methyl-n-2s,3r,4r,5r-2,3,4,5,6-pentahydroxyhexyl octanamide,octanoyl-n-methylglucamide PubChem CID: 160091 IUPAC Name: N-methyl-N-[(2S,3R,4R,5R)-2,3,4,5,6-pentahydroxyhexyl]octanamide SMILES: CCCCCCCC(=O)N(C)CC(C(C(C(CO)O)O)O)O