accessibility menu, dialog, popup

UserName

CAS RN 85259-19-4

H3COOClBr

CAS RN 85259-19-4

IUPAC Name: methyl 2-bromo-2-(2-chlorophenyl)acetate
Synonyms: methyl 2-bromo-2-(2-chlorophenyl)acetate
Molecular Weight (g/mol): 263.52
Molecular Formula: C9H8BrClO2
InChi Key: HMBUCZUZRQQJQD-UHFFFAOYNA-N
SMILES: COC(=O)C(Br)C1=CC=CC=C1Cl
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results