accessibility menu, dialog, popup

UserName

H2NFF

CAS RN 85118-06-5

IUPAC Name: 1-(2,5-difluorophenyl)methanamine
Synonyms: 1-(2,5-difluorophenyl)methanamine
Molecular Weight (g/mol): 143.14
Molecular Formula: C7H7F2N
InChi Key: GDFBHCMFIUBEQT-UHFFFAOYSA-N
SMILES: NCC1=CC(F)=CC=C1F
Boiling Point 
  • (3)
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (4)
  • (3)
  • (1)
  • (1)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)