accessibility menu, dialog, popup

UserName

CAS RN 85068-32-2

IUPAC Name: 2-[3,5-bis(trifluoromethyl)phenyl]acetonitrile
Synonyms: 2-[3,5-bis(trifluoromethyl)phenyl]acetonitrile
Molecular Weight (g/mol): 253.15
Molecular Formula: C10H5F6N
InChi Key: YXGWYBUKRTYHJM-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC(=CC(CC#N)=C1)C(F)(F)F
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results