accessibility menu, dialog, popup

UserName

CAS RN 850589-44-5

IUPAC Name: [3-chloro-4-(cyclopropylcarbamoyl)phenyl]boronic acid
Synonyms: 3-chloro-4-(cyclopropylcarbamoyl)phenylboronic acid
Molecular Weight (g/mol): 239.46
Molecular Formula: C10H11BClNO3
InChi Key: TUPWHDSMIIRKLY-UHFFFAOYSA-N
SMILES: OB(O)C1=CC(Cl)=C(C=C1)C(=O)NC1CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results