CAS RN 85-73-4
IUPAC Name:
2-({4-[(1,3-thiazol-2-yl)sulfamoyl]phenyl}carbamoyl)benzoic acid
Synonyms:
phthalylsulfathiazole phthalazole sulfaphthalazole thalistatyl thalazole phthalidin phthalylnorsulfazole ftalil-esteve ftalazol sulphaphthalyl
Molecular Weight (g/mol):
403.43
Molecular Formula:
C17H13N3O5S2
InChi Key:
PBMSWVPMRUJMPE-UHFFFAOYSA-N
SMILES:
OC(=O)C1=CC=CC=C1C(=O)NC1=CC=C(C=C1)S(=O)(=O)NC1=NC=CS1
1
–
1
of
1
results
1
Phthalylsulfathiazole 95.0+%, TCI America™
CAS: 85-73-4 Molecular Formula: C17H13N3O5S2 Molecular Weight (g/mol): 403.427 MDL Number: MFCD00005318 InChI Key: PBMSWVPMRUJMPE-UHFFFAOYSA-N PubChem CID: 4806 ChEBI: CHEBI:9336 IUPAC Name: 2-[[4-(1,3-thiazol-2-ylsulfamoyl)phenyl]carbamoyl]benzoic acid SMILES: C1=CC=C(C(=C1)C(=O)NC2=CC=C(C=C2)S(=O)(=O)NC3=NC=CS3)C(=O)O