accessibility menu, dialog, popup

UserName

CH3CH3

CAS RN 84540-37-4

IUPAC Name: 4-ethyl-4'-(4-propylcyclohexyl)-1,1'-biphenyl
Synonyms: 4-ethyl-4'-(4-propylcyclohexyl)-1,1'-biphenyl
Molecular Weight (g/mol): 306.49
Molecular Formula: C23H30
InChi Key: DOALOUQODWWGEZ-UHFFFAOYSA-N
SMILES: CCCC1CCC(CC1)C1=CC=C(C=C1)C1=CC=C(CC)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results