CAS RN 84504-69-8
CAS RN 84504-69-8
IUPAC Name:
(2Z)-but-2-enedioic acid; 6-(2,5-dichlorophenyl)-1,3,5-triazine-2,4-diamine
Synonyms:
irsogladine
Molecular Weight (g/mol):
372.16
Molecular Formula:
C13H11Cl2N5O4
InChi Key:
PJLVTVAIERNDEQ-BTJKTKAUSA-N
SMILES:
OC(=O)\C=C/C(O)=O.NC1=NC(=NC(N)=N1)C1=CC(Cl)=CC=C1Cl
1
–
1
of
1
results
1
Irsogladine Maleate 98.0+%, TCI America™
CAS: 84504-69-8 Molecular Formula: C13H11Cl2N5O4 Molecular Weight (g/mol): 372.16 MDL Number: MFCD00873566 InChI Key: PJLVTVAIERNDEQ-BTJKTKAUSA-N Synonym: irsogladine maleate,gaslon,gaslon n,unii-66gvu60epq,66gvu60epq,2,4-diamino-6-2,5-dichlorophenyl-s-triazine maleate,6-2,5-dichlorophenyl-1,3,5-triazine-2,4-diamine maleate,1,3,5-triazine-2,4-diamine, 6-2,5-dichlorophenyl-, z-2-butenedioate 1:1,dsstox_cid_20234 PubChem CID: 5282435 IUPAC Name: (2Z)-but-2-enedioic acid; 6-(2,5-dichlorophenyl)-1,3,5-triazine-2,4-diamine SMILES: OC(=O)\C=C/C(O)=O.NC1=NC(=NC(N)=N1)C1=CC(Cl)=CC=C1Cl