CAS RN 84501-67-7
IUPAC Name:
1-(2-bromoethyl)-4-ethyl-4,5-dihydro-1H-1,2,3,4-tetrazol-5-one
Synonyms:
1-(2-bromoethyl)-4-ethyl-1,2,3,4-tetrazol-5-one
Molecular Weight (g/mol):
221.06
Molecular Formula:
C5H9BrN4O
InChi Key:
KQKSCWCKLKLXJS-UHFFFAOYSA-N
SMILES:
CCN1N=NN(CCBr)C1=O
1
–
1
of
1
results
1
Promotions Available
1-(2-Bromoethyl)-4-ethyl-1,4-dihydro-5H-tetrazol-5-one, 95%
1-(2-Bromoethyl)-4-ethyl-1,4-dihydro-5H-tetrazol-5-one, 95%
CAS: 84501-67-7 Molecular Formula: C5H9BrN4O Molecular Weight (g/mol): 221.058 MDL Number: MFCD16251259 InChI Key: KQKSCWCKLKLXJS-UHFFFAOYSA-N Synonym: 1-2-bromoethyl-4-ethyl-1,4-dihydro-5h-tetrazol-5-one,1-2-bromoethyl-4-ethyltetrazol-5-one,1-2-bromoethyl-4-ethyl-tetrazol-5-one,1-2-bromoethyl-4-ethyl-1,2,3,4-tetrazol-5-one,2-4-ethyl-4,5-dihydro-5-oxo-1h-tetrazol-1-yl ethyl bromide PubChem CID: 3019915 IUPAC Name: 1-(2-bromoethyl)-4-ethyltetrazol-5-one SMILES: CCN1C(=O)N(N=N1)CCBr