accessibility menu, dialog, popup

UserName

H3COSOONOO

CAS RN 84271-78-3

IUPAC Name: 2,5-dioxopyrrolidin-1-yl 3-(acetylsulfanyl)propanoate
Synonyms: 2,5-dioxopyrrolidin-1-yl 3-(acetylsulfanyl)propanoate
Molecular Weight (g/mol): 245.25
Molecular Formula: C9H11NO5S
InChi Key: ZRTJVRDXVSDKPX-UHFFFAOYSA-N
SMILES: CC(=O)SCCC(=O)ON1C(=O)CCC1=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results