accessibility menu, dialog, popup

UserName

CH3NNOClClSOHOO

CAS RN 84-57-1

IUPAC Name: 2,5-dichloro-4-(3-methyl-5-oxo-4,5-dihydro-1H-pyrazol-1-yl)benzene-1-sulfonic acid
Synonyms: 2,5-dichloro-4-(3-methyl-5-oxo-4H-pyrazol-1-yl)benzenesulfonic acid
Molecular Weight (g/mol): 323.14
Molecular Formula: C10H8Cl2N2O4S
InChi Key: KCEIVWKDBLAQKL-UHFFFAOYSA-N
SMILES: CC1=NN(C(=O)C1)C1=CC(Cl)=C(C=C1Cl)S(O)(=O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results