accessibility menu, dialog, popup

UserName

CAS RN 839-90-7

IUPAC Name: tris(2-hydroxyethyl)-1,3,5-triazinane-2,4,6-trione
Synonyms: tris(2-hydroxyethyl)-1,3,5-triazinane-2,4,6-trione
Molecular Weight (g/mol): 261.23
Molecular Formula: C9H15N3O6
InChi Key: BPXVHIRIPLPOPT-UHFFFAOYSA-N
SMILES: OCCN1C(=O)N(CCO)C(=O)N(CCO)C1=O
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)