accessibility menu, dialog, popup

UserName

CH3OOOFFF

CAS RN 83643-84-9

IUPAC Name: methyl 4,4,4-trifluoro-3-oxobutanoate
Synonyms: methyl 4,4,4-trifluoro-3-oxobutanoate
Molecular Weight (g/mol): 170.09
Molecular Formula: C5H5F3O3
InChi Key: LKMUBWWZTSZGGV-UHFFFAOYSA-N
SMILES: COC(=O)CC(=O)C(F)(F)F
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)