missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4,4,4-Trifluoroacetoacetate 96.0+%, TCI America™

Catalog No. T12455G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T12455G 5 g
T124525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T12455G Supplier TCI America Supplier No. T12455G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 83643-84-9
Molecular Formula C5H5F3O3
Molecular Weight (g/mol) 170.087
MDL Number MFCD00041004
InChI Key LKMUBWWZTSZGGV-UHFFFAOYSA-N
Synonym methyl trifluoroacetoacetate, methyl 4,4,4-trifluoroacetoacetate, 4,4,4-trifluoroacetoacetic acid methyl ester, methyl-4,4,4-trifluoro-3-oxobutanoate, butanoic acid, 4,4,4-trifluoro-3-oxo-, methyl ester, acmc-209psv, methyltri-fluoroacetoacetate, methyl4,4,4-trifluoroacetoacetate, methyl-4,4,4-trifluoroacetoacetate, methyl 3-oxo-4,4,4-trifluorobutyrate
PubChem CID 550266
IUPAC Name methyl 4,4,4-trifluoro-3-oxobutanoate
SMILES COC(=O)CC(=O)C(F)(F)F

Specifications

Color Yellow
Boiling Point 116°C
Quantity 5 g
UN Number 3272
Formula Weight 170.09
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl 4,4,4-Trifluoroacetoacetate